7-Amino-2H-chromen-2-one 97%

Supplier: Ambeed
Warning

A689142-100MG A689142-1G A689142-250MG
76697-246EA 86 USD
76697-246 76697-248 76697-250
7-Amino-2H-chromen-2-one 97%
7-Amino-2H-chromen-2-one

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MDL Number: MFCD07367608
CAS Number: 19063-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR