Boc-d-Asp(ochex)-OH 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-D-aspartic acid 4-cyclohexyl ester

A688354-1G A688354-25G
76697-234EA 17 USD
76697-234 76697-236
Boc-d-Asp(ochex)-OH 98%
Boc-d-Asp(ochex)-OH

Precaución: for Research Use Only

Formula: C₁₅H₂₅NO₆
MDL Number: MFCD00060741
CAS Number: 112898-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR