Allylbenzene 98%

Supplier: Ambeed
Danger

Synonyms: 1-Phenyl-2-propene, 3-Phenylpropene

A687141-5G A687141-25G
76697-232EA 7.43 USD
76697-232 77552-088
Allylbenzene 98%
Allylbenzene

Precaución: for Research Use Only

Formula: (C₆H₅)CH₂CHCH₂
MW: 118.18 g/mol
MDL Number: MFCD00008651
CAS Number: 300-57-2
UN: 3295
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR