Dibenzo[b,e][1,4]dioxine 97%

Supplier: Ambeed
Warning

Synonyms: Dibenzo-p-dioxine

A684876-250MG A684876-1G A684876-5G
76697-228EA 63 USD
76697-228 77880-422 77880-423
Dibenzo[b,e][1,4]dioxine 97%
Dibenzo[b,e][1,4]dioxine

Precaución: for Research Use Only

Formula: C₁₂H₈O₂
MDL Number: MFCD00022282
CAS Number: 262-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR