Cocarboxylase 98%

Supplier: Ambeed
Warning

Synonyms: Thiamine pyrophosphate chloride, Aneurinepyrophosphoric acid, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-((hydroxy(phosphonooxy)phosphoryl)oxy)ethyl)-4-methylthiazol-3-ium chloride, Cocarboxylase chloride

A681169-100G A681169-1G A681169-25G A681169-5G
76697-212EA 935.72 USD
76697-212 76697-214 76697-216 76697-218
Cocarboxylase 98%
Cocarboxylase

Precaución: for Research Use Only

Formula: C₁₂H₁₉ClN₄O₇P₂S
MW: 460.77 g/mol
MDL Number: MFCD00038740
CAS Number: 154-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR