Acid red 1 90-100% (dye content)

Supplier: Ambeed
Warning

Synonyms: Red 2G, C.I. 18050, Amido Naphthol Red G, Sodium 5-acetamido-4-hydroxy-3-(phenyldiazenyl)naphthalene-2,7-disulphonate, 5-Acetamido-4-hydroxy-3-phenylazo-2,7-naphthalene-disulphonic acid disodium salt, Pontacyl Carmine, Azophloxine

A680914-1000G A680914-100G A680914-500G
76697-206EA 81 USD
76697-206 76697-208 76697-210
Acid red 1 90-100% (dye content)
Acid red 1

Precaución: for Research Use Only

Formula: C₁₈H₁₅N₃O₈S₂
MDL Number: MFCD00003954
CAS Number: 3734-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR