Copper(II) acetate 98%
Supplier: Ambeed
|
Danger
|
A680709-10G
A680709-25G
A680709-100G
A680709-500G
76697-202EA
12
USD
76697-202
76697-204
77552-070
77573-206
Copper(II) acetate 98%
Copper(II) acetate
Precaución: for Research Use Only
Formula:
CuCH₃CO₂ |
MDL Number:
MFCD00008690 CAS Number: 142-71-2 UN: 1759 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...