Coumarin 153 98%

Supplier: Ambeed
Warning

Synonyms: Coumarin 540A, 2,3,6,7-Tetrahydro-9-(trifluoromethyl)-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizin-11-one, Coumarin153

A680384-100MG A680384-1G A680384-250MG
76697-196EA 88.06 USD
76697-196 76697-198 76697-200
Coumarin 153 98%
Coumarin 153

Precaución: for Research Use Only

Formula: C₁₆H₁₄F₃NO₂
MW: 309.29 g/mol
MDL Number: MFCD00041843
CAS Number: 53518-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR