(Triphenylphosphine)gold(I) bromide 98%

Supplier: Ambeed
Warning

Synonyms: Bromo(triphenylphosphine)gold(I)

A678716-100MG A678716-250MG A678716-1G
76697-184EA 19 USD
76697-184 76697-186 77878-515
(Triphenylphosphine)gold(I) bromide 98%
(Triphenylphosphine)gold (I) bromide

Precaución: for Research Use Only

Formula: C₁₈H₁₅AuBrP
MW: 539.16
MDL Number: MFCD00049267
CAS Number: 14243-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR