N-Carbobenzoxy-1,6-diaminohexane hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: N-Cbz-1,6-diaminohexane Hydrochloride, Benzyl N-(6-Aminohexyl)carbamate Hydrochloride, Benzyl (6-aminohexyl)carbamate hydrochloride, N-(6-Aminohexyl)carbamic Acid Benzyl Ester Hydrochloride

A676299-1G A676299-250MG A676299-5G A676299-25G
76697-170EA 84 USD
76697-170 77627-370 77627-372 77878-510
N-Carbobenzoxy-1,6-diaminohexane hydrochloride 95%
N-Carbobenzoxy-1,6-diaminohexane hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₂₂N₂O₂
MW: 286.80 g/mol
MDL Number: MFCD00270151
CAS Number: 78618-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR