Bis(1,2,2,6,6-pentamethyl-4-piperidyl)sebacate 90%

Supplier: Ambeed
Warning

Synonyms: Bis(1,2,2,6,6-pentamethylpiperidin-4-yl) decanedioate, Decanedioic Acid Bis(1,2,2,6,6-pentamethyl-4-piperidyl) Ester, Sebacic Acid Bis(1,2,2,6,6-pentamethyl-4-piperidyl) Ester

A675594-1000G A675594-100G A675594-10G A675594-25G A675594-500G
76697-160EA 425 USD
76697-160 76697-162 76697-164 76697-166 76697-168
Bis(1,2,2,6,6-pentamethyl-4-piperidyl)sebacate 90%
Bis(1,2,2,6,6-pentamethyl-4-piperidyl)sebacate

Precaución: for Research Use Only

Formula: C₃₀H₅₆N₂O₄
MW: 508.79 g/mol
MDL Number: MFCD00134706
CAS Number: 41556-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR