Abacavir sulfate 98%

Supplier: Ambeed
Danger

Synonyms: ((1S,4R)-4-(2-Amino-6-(cyclopropylamino)-9H-purin-9-yl)cyclopent-2-en-1-yl)methanol hemisulphate

A674299-10MG A674299-50MG A674299-1G A674299-250MG A674299-5MG A674299-100MG A674299-1MG
76697-150EA 5.31 USD
76697-150 76697-152 77602-714 77627-360 77627-362 77627-356 77627-358
Abacavir sulfate 98%
Abacavir sulfate

Precaución: for Research Use Only

Formula: C₂₈H₃₈N₁₂O₆S
MW: 670.74 g/mol
MDL Number: MFCD04112763
CAS Number: 188062-50-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR