7,7-Dichlorobicyclo[3.2.0]hept-2-en-6-one 98%

Supplier: Ambeed
Danger

A672749-1G A672749-25G
76697-146EA 36.08 USD
76697-146 76697-148
7,7-Dichlorobicyclo[3.2.0]hept-2-en-6-one 98%
7,7-Dichlorobicyclo[3.2.0]hept-2-en-6-one

Precaución: for Research Use Only

Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
MDL Number: MFCD00013738
CAS Number: 5307-99-3
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR