(6R,6aR,13R,13aS)-2,3,4,6,6a,7,8,9,10,12,13,13a-Dodecahydro-1H-6,13-methanodipyrido[1,2-a:3',2'-e]azocine 98%

Supplier: Ambeed
Warning

Synonyms: Aloperine

A672020-1G A672020-1MG A672020-250MG
76697-134EA 180.36 USD
76697-134 76697-136 76697-138
(6R,6aR,13R,13aS)-2,3,4,6,6a,7,8,9,10,12,13,13a-Dodecahydro-1H-6,13-methanodipyrido[1,2-a:3',2'-e]azocine 98%
(6R,6aR,13R,13aS)-2,3,4,6,6a,7,8,9,10,12,13,13a-Dodecahydro-1H-6,13-methanodipyrido[1,2-a:3',2'-e]azocine

Precaución: for Research Use Only

Formula: C₁₅H₂₄N₂
MW: 232.36 g/mol
MDL Number: MFCD00468086
CAS Number: 56293-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR