9-Decyn-1-ol 90%

Supplier: Ambeed
Warning

A671482-1G A671482-250MG A671482-25G A671482-5G A671482-100G
76697-126EA 41 USD
76697-126 76697-128 76697-130 76697-132 77627-346
9-Decyn-1-ol 90%
9-Decyn-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₈O
MDL Number: MFCD00049199
CAS Number: 17643-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR