2,4,6-Trimethylbenzenesulfonamide 96%

Supplier: Ambeed
Warning

A671278-1G A671278-25G A671278-5G
76697-120EA 17 USD
76697-120 76697-122 76697-124
2,4,6-Trimethylbenzenesulfonamide 96%
2,4,6-Trimethylbenzenesulphonamide

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂S
MDL Number: MFCD00463245
CAS Number: 4543-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR