Clonixin 98%

Supplier: Ambeed
Warning

Synonyms: 2-[(3-Chloro-2-methylphenyl)amino]nicotinic Acid, 2-(3-Chloro-2-methylanilino)pyridine-3-carboxylic acid, 2-(3-Chloro-o-toluidino)nicotinic Acid

A668370-100MG A668370-250MG A668370-50MG A668370-5G A668370-10MG
76697-112EA 7.43 USD
76697-112 76697-114 76697-116 76697-118 77627-340
Clonixin 98%
Clonixin

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClN₂O₂
MW: 262.69 g/mol
MDL Number: MFCD00864292
CAS Number: 17737-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR