1,1'-(Decane-1,10-diyl)bis(4-amino-2-methylquinolin-1-ium) chloride 98%

Supplier: Ambeed
Warning

Synonyms: Dequalinium Chloride

A666741-100MG A666741-250MG
76697-104EA 47.75 USD
76697-104 76697-106
1,1'-(Decane-1,10-diyl)bis(4-amino-2-methylquinolin-1-ium) chloride 98%
1,1'-(Decane-1,10-diyl)bis(4-amino-2-methylquinolin-1-ium) chloride

Precaución: for Research Use Only

Formula: C₃₀H₄₀Cl₂N₄
MW: 527.57 g/mol
MDL Number: MFCD00063502
CAS Number: 522-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR