4-Amino-N-(1-benzylpiperidin-4-yl)-5-chloro-2-methoxybenzamide 2-hydroxysuccinate 98%

Supplier: Ambeed
Warning

A666579-10MG A666579-1MG A666579-50MG A666579-5MG
76697-096EA 65 USD
76697-096 76697-098 76697-100 76697-102
4-Amino-N-(1-benzylpiperidin-4-yl)-5-chloro-2-methoxybenzamide 2-hydroxysuccinate 98%
4-Amino-N-(1-benzylpiperidin-4-yl)-5-chloro-2-methoxybenzamide 2-hydroxysuccinate

Precaución: for Research Use Only

Formula: C₂₄H₃₀ClN₃O₇
MDL Number: MFCD01743960
CAS Number: 57645-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR