(E)-3-([1,1'-Biphenyl]-4-yl)acrylic acid 97%

Supplier: Ambeed
Danger

A666232-1G A666232-25G
76697-094EA 11 USD
76697-094 77880-396
(E)-3-([1,1'-Biphenyl]-4-yl)acrylic acid 97%
(E)-3-([1,1'-Biphenyl]-4-yl)acrylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₂O₂
MDL Number: MFCD00014010
CAS Number: 88241-65-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR