Aluminium hydroxide 99%

Supplier: Ambeed
Warning

A663451-1000G A663451-25MG A663451-500G A663451-100G
76697-078EA 10.61 USD
76697-078 76697-080 76697-082 77627-332
Aluminium hydroxide 99%
Aluminium hydroxide

Precaución: for Research Use Only

Formula: Al(OH)₃
MDL Number: MFCD00003420
CAS Number: 21645-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR