9-(Bromomethyl)acridine 98%

Supplier: Ambeed
Danger

A663408-100MG A663408-1G A663408-250MG A663408-5G
76697-072EA 11 USD
76697-072 76697-074 76697-076 77552-024
9-(Bromomethyl)acridine 98%
9-(Bromomethyl)acridine

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrN
MDL Number: MFCD01321146
CAS Number: 1556-34-9
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR