2,4,5-Trimethylbenzoic acid 98%

Supplier: Ambeed
Warning

A662591-100G A662591-10G A662591-1G A662591-25G
76697-062EA 781.89 USD
76697-062 76697-064 76697-066 76697-068
2,4,5-Trimethylbenzoic acid 98%
2,4,5-Trimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
MDL Number: MFCD00075863
CAS Number: 528-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR