2,5-Dimethoxybenzhydrazide 95%

Supplier: Ambeed
Warning

A662055-1G A662055-250MG A662055-5G
76697-056EA 20 USD
76697-056 76697-058 76697-060
2,5-Dimethoxybenzhydrazide 95%
2,5-Dimethoxybenzhydrazide

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₃
MDL Number: MFCD00017049
CAS Number: 17894-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR