(+)-Dimethyl-D-malate 98%

Supplier: Ambeed
Danger

Synonyms: D-(+)-Malic acid dimethyl ester, Dimethyl D-(+)-Malate, D-(+)-Apple acid dimethyl ester, (R)-Dimethyl 2-hydroxysuccinate

A660803-25G A660803-5G
76697-044EA 110 USD
76697-044 76697-046
(+)-Dimethyl-D-malate 98%
(+)-Dimethyl-D-malate

Precaución: for Research Use Only

Formula: C₆H₁₀O₅
MDL Number: MFCD00066216
CAS Number: 70681-41-3
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR