Allyl furan-2-carboxylate 98% stabilized

Supplier: Ambeed
Warning

Synonyms: Allyl 2-furoate

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A658779-100G A658779-1G A658779-25G A658779-5G
76697-028EA 248 USD
76697-028 76697-030 76697-032 76697-034
Allyl furan-2-carboxylate 98% stabilized
Allyl furan-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₈O₃
MW: 152.14 g/mol
MDL Number: MFCD00016580
CAS Number: 4208-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR