4-Ethoxy-3-methoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A657116-10G A657116-1G A657116-25G A657116-5G A657116-100G
76697-010EA 15 USD
76697-010 76697-012 76697-014 76697-016 77880-380
4-Ethoxy-3-methoxybenzaldehyde 98%
4-Ethoxy-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MDL Number: MFCD00016614
CAS Number: 120-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR