4-(4-Fluorophenyl)butyric acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Fluorophenyl)butyric Acid

A652989-1G A652989-25G A652989-5G
76696-990EA 46.68 USD
76696-990 76696-992 76696-994
4-(4-Fluorophenyl)butyric acid 97%
4-(4-Fluorophenyl)butyric acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂
MW: 182.2 g/mol
MDL Number: MFCD03788503
CAS Number: 589-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR