Copper(II)-2-ethylhexanoate Cu ~16.5-19%

Supplier: Ambeed
Warning

Synonyms: 2-Ethylhexanoic acid copper (II) salt, Copper(II) 2-ethylhexanoate, 2-Ethylcaproic acid copper (II) salt, Copper bis(2-ethylhexanoate)

A652646-1G A652646-25G A652646-5G A652646-10G
76696-984EA 8.49 USD
76696-984 76696-986 76696-988 77602-666
Copper(II)-2-ethylhexanoate Cu ~16.5-19%
Copper(II)-2-ethylhexanoate

Precaución: for Research Use Only

Formula: C₁₆H₃₀CuO₄
MW: 349.96 g/mol
MDL Number: MFCD00015695
CAS Number: 149-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR