Bromocresol purple sodium salt indicator

Supplier: Ambeed
Warning

Synonyms: Sodium α-(3-bromo-5-methyl-4-oxo-2,5-cyclohexadienylidene)-α-(3-bromo-5-methyl-4-hydroxyphenyl)toluenesulphonate, Bromocresol purple water soluble, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-methylphenolate

A647791-5G A647791-100G A647791-25G
76696-976EA 11 USD
76696-976 77878-470 77878-471
Bromocresol purple sodium salt indicator
Bromocresol purple sodium salt

Precaución: for Research Use Only

Formula: C₂₁H₁₅Br₂NaO₅S
MDL Number: MFCD00148896
CAS Number: 62625-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR