(2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-2-(((3S,4R,4aS)-8-oxo-4-vinyl-3,4,4a,5,6,8-hexahydropyrano[3,4-c]pyran-3-yl)oxy)tetrahydro-2H-pyran-3-yl 3,3',5-trihydroxy-[1,1'-biphenyl]-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Amarogentin

A647580-10MG A647580-1MG A647580-25MG A647580-50MG A647580-5MG
76696-966EA 61.54 USD
76696-966 76696-968 76696-970 76696-972 76696-974
(2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-2-(((3S,4R,4aS)-8-oxo-4-vinyl-3,4,4a,5,6,8-hexahydropyrano[3,4-c]pyran-3-yl)oxy)tetrahydro-2H-pyran-3-yl 3,3',5-trihydroxy-[1,1'-biphenyl]-2-carboxylate 97%
(2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-2-(((3S,4R,4aS)-8-oxo-4-vinyl-3,4,4a,5,6,8-hexahydropyrano[3,4-c]pyran-3-yl)oxy)tetrahydro-2H-pyran-3-yl 3,3',5-trihydroxy-[1,1'-biphenyl]-2-carboxylate

Precaución: for Research Use Only

Formula: C₂₉H₃₀O₁₃
MW: 586.54 g/mol
MDL Number: MFCD06656289
CAS Number: 21018-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR