Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate 97%

Supplier: Ambeed
Warning

Synonyms: Bis(1,5-cyclooctadiene)iridium (I) tetrafluoroborate

A647090-100MG A647090-1G A647090-5G
76696-956EA 47.75 USD
76696-956 76696-958 76696-960
Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate 97%
Bis(1,5-cyclooctadiene)iridium (I) tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₆H₂₄BF₄Ir
MW: 495.383 g/mol
MDL Number: MFCD00079664
CAS Number: 35138-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR