Dimethyl-4-aminothiophene-2,3-dicarboxylate hydrochloride 97%

Supplier: Ambeed
Warning

A645242-100MG A645242-5G
76696-940EA 11 USD
76696-940 77878-456
Dimethyl-4-aminothiophene-2,3-dicarboxylate hydrochloride 97%
Dimethyl-4-aminothiophene-2,3-dicarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO₄S
MDL Number: MFCD00205189
CAS Number: 121071-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR