cis-3-Hexen-1-yltiglate 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tiglic Acid cis-3-Hexen-1-yl Ester, (E)-(Z)-Hex-3-en-1-yl 2-methylbut-2-enoate
A643920-100G
A643920-1G
A643920-25G
A643920-5G
76696-932EA
183.54
USD
76696-932
76696-934
76696-936
76696-938
cis-3-Hexen-1-yltiglate 98+%
cis-3-Hexen-1-yltiglate
Precaución: for Research Use Only
Formula:
C₁₁H₁₈O₂ MW: 182.26 g/mol |
MDL Number:
MFCD00036528 CAS Number: 67883-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...