1-Isopropoxy-4-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: Isopropyl 4-Nitrophenyl Ether

A643785-100G A643785-1G A643785-25G A643785-5G
76696-924EA 374.5 USD
76696-924 76696-926 76696-928 76696-930
1-Isopropoxy-4-nitrobenzene 98%
1-Isopropoxy-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
MDL Number: MFCD01327671
CAS Number: 26455-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR