Bis(2-bromobenzyl)amine hydrochloride 95%

Supplier: Ambeed
Warning

A643659-1G A643659-5G
76696-916EA 28.65 USD
76696-916 76696-918
Bis(2-bromobenzyl)amine hydrochloride 95%
Bis(2-bromobenzyl)amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₁₃Br₂N·HCl
MW: 391.53 g/mol
MDL Number: MFCD01632133
CAS Number: 336615-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR