Barium stearate 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
Stearic acid barium salt
A643051-1000G
A643051-100G
A643051-500G
76696-910EA
20
USD
76696-910
76696-912
76696-914
Barium stearate 98+%
Barium stearate
Precaución: for Research Use Only
Formula:
C₃₆H₇₀BaO₄ MW: 704.28 g/mol Melting Pt: 160 |
MDL Number:
MFCD00050711 CAS Number: 6865-35-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...