4-Methyl-2-phenyl-1,3-dioxolane (mixture of isomers) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Benzaldehyde Propylene Glycol Acetal
A642754-100G
A642754-25G
76696-906EA
37.14
USD
76696-906
76696-908
4-Methyl-2-phenyl-1,3-dioxolane (mixture of isomers) 98%
4-Methyl-2-phenyl-1,3-dioxolane (mixture of isomers)
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₂ MW: 164.20 g/mol |
MDL Number:
MFCD00059732 CAS Number: 2568-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...