Boc-Met(O2)-OH 95%

Supplier: Ambeed
Warning

Synonyms: Boc-Met(O₂)-OH

A642473-1G A642473-100G A642473-25G A642473-5G
76696-904EA 17 USD
76696-904 77627-282 77627-284 77627-286
Boc-Met(O2)-OH 95%
Boc-Met(O2)-OH

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₆S
MDL Number: MFCD00065587
CAS Number: 60280-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR