4,4',4''-(2-Chloroethene-1,1,2-triyl)tris(methoxybenzene) 98%

Supplier: Ambeed
Warning

Synonyms: Chlorotrianisene

A640644-1MG A640644-25MG
76696-894EA 8.49 USD
76696-894 76696-896
4,4',4''-(2-Chloroethene-1,1,2-triyl)tris(methoxybenzene) 98%
4,4',4''-(2-Chloroethene-1,1,2-triyl)tris(methoxybenzene)

Precaución: for Research Use Only

Formula: C₂₃H₂₁ClO₃
MW: 380.86 g/mol
MDL Number: MFCD00048044
CAS Number: 569-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR