N-[(S)-1-Carbethoxy-1-butyl]-L-alanine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(1-Carbethoxy-1-butyl)-Ala-OH
A640403-100G
A640403-1G
A640403-25G
A640403-5G
76696-884EA
265
USD
76696-884
76696-886
76696-888
76696-890
N-[(S)-1-Carbethoxy-1-butyl]-L-alanine 95%
N-[(S)-1-Carbethoxy-1-butyl]-L-alanine
Precaución: for Research Use Only
Formula:
C₁₀H₁₉NO₄ |
MDL Number:
MFCD07782126 CAS Number: 82834-12-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...