6-(Trifluoromethyl)pyridine-3-amidine hydrochloride 99%

Supplier: Ambeed
Danger

Synonyms: 6-(Trifluoromethyl)nicotinimidamide hydrochloride, 6-(Trifluoromethyl)nicotinamidine hydrochloride

A638344-1G A638344-250MG
76696-868EA 45.62 USD
76696-868 76696-870
6-(Trifluoromethyl)pyridine-3-amidine hydrochloride 99%
6-(Trifluoromethyl)pyridine-3-amidine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₇ClF₃N₃
MW: 225.6 g/mol
MDL Number: MFCD01862652
CAS Number: 221313-11-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR