7-Bromo-5-chloro-8-hydroxyquinoline 98%

Supplier: Ambeed
Warning

A635486-1G A635486-250MG A635486-5G
76696-862EA 26 USD
76696-862 76696-864 76696-866
7-Bromo-5-chloro-8-hydroxyquinoline 98%
7-Bromo-5-chloro-8-hydroxyquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrClNO
MDL Number: MFCD00044601
CAS Number: 7640-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR