Allyl(chloromethyl)dimethylsilane 97+% stabilized

Supplier: Ambeed
Danger

Synonyms: 3-Chloromethyldimethylsilyl-1-propene

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A634612-1G A634612-5G
76696-858EA 13.8 USD
76696-858 76696-860
Allyl(chloromethyl)dimethylsilane 97+% stabilized
Allyl(chloromethyl)dimethylsilane

Precaución: for Research Use Only

Formula: C₆H₁₃ClSi
MW: 148.71 g/mol
MDL Number: MFCD00012045
CAS Number: 75422-66-1
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR