(2,4-Dimethoxyphenyl)acetic acid 98%

Supplier: Ambeed
Warning

A633324-1G A633324-25G A633324-5G
76696-834EA 20 USD
76696-834 76696-836 76696-838
(2,4-Dimethoxyphenyl)acetic acid 98%
(2,4-Dimethoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃(OCH₃)₂)CH₂COOH
MDL Number: MFCD00183328
CAS Number: 6496-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR