Bis(pentamethylcyclopentadienyl)nickel(II) 98%

Supplier: Ambeed
Danger

A632728-100MG A632728-1G A632728-250MG A632728-5G
76696-818EA 46 USD
76696-818 76696-820 77627-266 77878-444
Bis(pentamethylcyclopentadienyl)nickel(II) 98%
Bis(pentamethylcyclopentadienyl)nickel (II)

Precaución: for Research Use Only

Formula: C₂₀H₃₀Ni
MDL Number: MFCD00058704
CAS Number: 74507-63-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR