Alizarine yellow GG (sodium salt) indicator

Supplier: Ambeed
Warning

Synonyms: Salicyl Yellow, Sodium 2-hydroxy-5-((3-nitrophenyl)diazenyl)benzoate, Metachrome Yellow Mordant Yellow 1, Alizarin Yellow 2G, 5-(3-Nitrophenylazo)salicylic acid sodium salt, Metachrome Yellow, Alizarin yellow GG

A632278-100G A632278-25G A632278-500G
76696-812EA 48 USD
76696-812 76696-814 76696-816
Alizarine yellow GG (sodium salt) indicator
Alizarine yellow GG (sodium salt)

Precaución: for Research Use Only

Formula: C₁₃H₈N₃NaO₅
MDL Number: MFCD00044746
CAS Number: 584-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR