6-Nitroquinolin-5-ylamine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-6-nitroquinoline, 6-Nitroquinolin-5-amine

A632253-1G A632253-250MG A632253-25G A632253-5G
76696-804EA 53.05 USD
76696-804 76696-806 76696-808 76696-810
6-Nitroquinolin-5-ylamine 97%
6-Nitroquinolin-5-ylamine

Precaución: for Research Use Only

Formula: C₉H₇N₃O₂
MW: 189.17 g/mol
MDL Number: MFCD00006798
CAS Number: 35975-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR