Benzyl (2-hydrazinyl-2-oxoethyl)carbamate 97%

Supplier: Ambeed
Warning

Synonyms: Cbz-Gly-OH Hydrazide

A631354-100MG A631354-1G A631354-250MG A631354-25G A631354-5G
76696-794EA 11 USD
76696-794 76696-796 76696-798 76696-800 77880-340
Benzyl (2-hydrazinyl-2-oxoethyl)carbamate 97%
Benzyl (2-hydrazinyl-2-oxoethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₃N₃O₃
MW: 223.23 g/mol
MDL Number: MFCD00025127
CAS Number: 5680-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR