α-Cyclopropylbenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: alpha-Cyclopropylbenzyl alcohol, Cyclopropyl(phenyl)methanol, α-Cyclopropylbenzyl alcohol, Cyclopropyl phenyl carbinol, Cyclopropylphenylcarbinol

A630001-1G A630001-25G A630001-5G
76696-782EA 39 USD
76696-782 76696-784 76696-786
α-Cyclopropylbenzyl alcohol 98%
α-Cyclopropylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MDL Number: MFCD00001299
CAS Number: 1007-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR